Oncomine™ TCR Beta-SR Assay, RNA
Thermo Scientific™

Oncomine™ TCR Beta-SR Assay, RNA

The Oncomine TCR Beta-SR Assays are robust, targeted next-generation sequencing (NGS) assays designed to accurately identify and measure the clonalRead more
Have Questions?
Catalog NumberQuantity
A3935924 Reactions
Catalog number A39359
Price (USD)
8,590.00
Each
Quantity:
24 Reactions
Price (USD)
8,590.00
Each

The Oncomine TCR Beta-SR Assays are robust, targeted next-generation sequencing (NGS) assays designed to accurately identify and measure the clonal expansion of T lymphocytes in formalin-fixed paraffin-embedded (FFPE) tumor samples. These assays identify unique T-cell clones through targeting of the diverse complementarity-determining region 3 (CDR3) of the T-cell receptor (TCR) gene locus in genomic DNA or RNA. The nucleotide sequence of the CDR3 region is unique to each T-cell clone and codes for the part of the TCR beta chain that is involved in antigen recognition. This particular assay is for use with RNA samples. For DNA samples, please see Cat. No. A39072.

Benefits of the Oncomine TCR Beta-SR Assay - RNA include:

  • Compatibility with a vast array of research sample types, including FFPE tissue, fresh-frozen (FF) tissue, whole blood, PBLs, and PBMCs
  • Low input requirement
  • Clonality data for only productive rearrangements
  • High sensitivity and low limit of detection (LoD) for rare clone identification through dual-barcode indexing
  • Efficient workflow with 48-hr sample-to-results time
  • Flexible input requirements ranging from 50 ng to 1 μg
  • Streamlined and user-friendly informatics solution with multi-sample analysis functionality

The Oncomine TCR Beta-SR Assay - RNA uses multiplex PCR primers to generate 80-bp amplicons that can be sequenced on all chip types supported by the Ion GeneStudio S5 sequencing systems, allowing you to pick the best multiplexing configuration for your unique sample batching needs and throughput requirements. The entire workflow (figure below) from isolation of RNA to analysis of samples can be accomplished in two days using the Ion Chef templating system and Ion GeneStudio S5 system. The assay kit provides a single pool of multiplex PCR primers and library reagents and is compatible with sample types such as RNA from FFPE tissue, fresh-frozen (FF) tissue, whole blood, PBLs, or PBMCs.

The Oncomine TCR Beta-SR Assay - RNA supports key applications in the field of immuno-oncology and hemato-oncology research. The high-sensitivity dual-barcode indexing, flexible input requirements (50 ng-1 μg), high-depth sequencing, and high-throughput capability make this assay ideal for minimal residual disease testing for T-cell malignancies in blood. The unique FFPE compatibility of this assay enables researchers to focus on T-cell infiltration and clonality in the tumor microenvironment. Measuring T-cells in the tumor microenvironment is useful in biomarker discovery and in monitoring an individual's response to immunotherapies such as checkpoint blockade inhibitors, cancer vaccines, or chimeric antigen receptor (CAR) T-cell therapies.

Learn more about the assay

Note: Information about data analysis using Ion Reporter Software v5.10 can be found in the User Guide below.

Specifications
For Use With (Application)Immune Repertoire Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of SamplesUp to 24 Samples
Nucleic Acid TypeRNA
Product LineOncomine
Product TypeAssay
Quantity24 Reactions
Sample TypeWhole Blood, PBMC, Bone Marrow, Fresh/Frozen Tissue, FFPE Tissue
ThroughputChip-dependent Multiplexing
Workflow StepPanel and Library
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.