Expi293™ PRO Expression Medium
Gibco™

Expi293™ PRO Expression Medium

Expi293 PRO Expression Medium supports rapid, high-yield protein production from HEK293 cells. Designed for scalable, high-density cultures with consistent expression across a variety of proteins, including complex and low-yield targets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A40001355056 x 1 L
A40001355011 L
A400013550310 L
A400013550420 L
Catalog number A4000135505
Price (INR)
138,910.00
Online offer
Ends: 30-Sep-2026
252,562.00
Save 113,652.00 (45%)
Each
Quantity:
6 x 1 L
Price (INR)
138,910.00
Online offer
Ends: 30-Sep-2026
252,562.00
Save 113,652.00 (45%)
Each

Gibco Expi293 PRO Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293 PRO cells in the Expi293 PRO Expression System for scalable transient protein expression.

  • Supports growth of suspension HEK 293 cultures to densities of over 15 x 10^6 cells/mL
  • Transfection-compatible serum-free suspension culture medium
  • Helps enable sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 5 g/L of recombinant protein
  • Supports growth and transfection of HEK293 cells in culture formats of 1 mL to 10 L
  • Does not contain phenol red

Expi293 PRO Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 PRO Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeEmbryonic Kidney Cells
Product LineExpi293 PRO
Product TypeExpression Medium
Quantity6 x 1 L
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesGlutaMAX
Without AdditivesNo HEPES, No Phenol Red
Unit SizeEach
Contents & Storage
Store in the dark at +2°C to +8°C