InnoviGene™ Suite Software
Applied Biosystems™

InnoviGene™ Suite Software

InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing & fragment analysis data generated by Applied Biosystems genetic analyzers.
製品番号(カタログ番号)製品タイプライセンス
A40004742Sanger Sequencing & Fragment Analysis Software1 Year
A40004750Fragment Analysis Software180 Days (Demo)
A40004747Fragment Analysis Software1 Year
A40004740Fragment Analysis Software3 Years
A40004752Fragment Analysis Software5 years
A40004743Fragment Analysis Software Renewal1 Year
A40004741Fragment Analysis Software Renewal3 Years
A40004739Fragment Analysis Software Renewal5 Years
A40004746Sanger Sequencing & Fragment Analysis Software180 Days (Demo)
A40004744Sanger Sequencing & Fragment Analysis Software3 Years
A40004749Sanger Sequencing & Fragment Analysis Software5 years
A40004748Sanger Sequencing & Fragment Analysis Software Renewal1 Year
A40004751Sanger Sequencing & Fragment Analysis Software Renewal3 Years
A40004745Sanger Sequencing & Fragment Analysis Software Renewal5 Years
A40001760Sequencing Software180 Days (Demo)
A40001762Sequencing Software1 Year
A40001765Sequencing Software3 Years
A40001763Sequencing Software5 Years
製品番号(カタログ番号) A40004742
価格(JPY)
-
製品タイプ:
Sanger Sequencing & Fragment Analysis Software
ライセンス:
1 Year

Applied Biosystems InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing and fragment analysis data generated by Applied Biosystems genetic analyzers. The software provides a streamlined workflow to guide you through uploading and analyzing the data and generating reports.

InnoviGene Suite includes SAE administrator console software with security, audit, and electronic signature (SAE) functions that support compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.

The software is delivered via a download link sent to you after purchase.

Applications within InnoviGene Suite

InnoviGene Suite allows you to create projects for use with other applications that are integrated into the software. These applications enable you to complete an end-to-end workflow using projects that contain related files. The applications available depend on the type of InnoviGene Suite purchased ('sequencing' or 'fragment analysis').

Sequence Quality Check application (Innovigene Suite for sequencing)—enables you to perform the following activities:

  • View the quality information for trace files
  • Change quality settings and reanalyze trace files
  • Perform comparative analysis across multiple trace files

Sequence Identity application (InnoviGene Suite for sequencing)—enables you to view the assembly of multiple files compared to a reference and view basecall mismatches. Specifically, you can perform the following activities with the application:

  • Basecalling, quality value assignment, and mixed base identification
  • Trimming low-quality ends from each sequence, if the setting is active
  • Identifying poor-quality samples
  • Assembling the samples against the reference sequence and generating a specimen consensus sequence
  • Reviewing the basecalling quality values and consensus quality values
  • Identifying mismatches by aligning and comparing specimen sequences to the reference sequence. A mismatch is a position in the specimen that does not match with the reference sequence. Mismatches include base changes, insertions, and deletions.
  • Assigning specimen status based on alignment with the reference sequence and coverage criteria
  • Editing basecalls in samples or the consensus
  • Generating reports and results for export in CSV format

STR Typing application (InnoviGene Suite for fragment analysis)—the STR Typing application is genotyping software that uses short tandem repeat (STR) data. It enables you to perform the following activities:

  • Analyze a mixture of DNA fragments, separated by size
  • Provide a profile of the separation
  • Calculate the sizes of the fragments
  • Determine the alleles present in fragment analysis sample (FSA) files
研究用途にのみご使用ください。診断目的には使用できません。
仕様
使用対象(アプリケーション)Sanger Sequencing & Fragment Analysis
使用対象 (装置)Applied Biosystems™ Genetic Analyzers
フォーマットSoftware
ライセンス1 Year
製品タイプSanger Sequencing & Fragment Analysis Software
数量5 Users
シーケンスタイプSanger Sequencing & Fragment Analysis
製品ラインInnoviGene
Unit SizeEach