New: Thermo Scientific™ Pierce™ Peptide Clean-Up Plates offer new sorbent chemistries in a 96-well format.
Pierce™ Peptide Clean-Up Plate, 96-well
Pierce™ Peptide Clean-Up Plate, 96-well
Pierce™ Peptide Clean-Up Plate, 96-well
Thermo Scientific™
Pierce™ Peptide Clean-Up Plate, 96-well
Thermo Scientific™ Pierce™ Peptide Clean-Up Plate products are sorbent-based 96-well plates designed for parallel peptide desalting, clean-up, concentration, and fractionation prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSorbent
A40012252C18
A40012250MCX
A40012254SAX
A40012255SDB-XC
A40012249SDB-RPS
Catalog number A40012252
Price (USD)
352.00
Each
Sorbent:
C18
Price (USD)
352.00
Each
Thermo Scientific™ Pierce™ Peptide Clean-Up Plate products are sorbent-based 96-well plates designed for parallel peptide desalting, clean-up, concentration, and fractionation prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows. These plates help remove salts, detergents, buffers, denaturants, chaotropes, and other chemistry-compatible contaminants that can interfere with downstream analysis. The plate portfolio includes multiple peptide clean-up chemistries, including octadecyl silica (C18), strong anion exchange (SAX), mixed-mode cation exchange (MCX), styrene divinylbenzene reversed-phase sulfonate (SDB-RPS), and styrene divinylbenzene crosslinked (SDB-XC), for high-throughput desalting, clean-up, concentration, and fractionation workflows.
Thermo Scientific™ Pierce™ Peptide Clean-Up Plate products use reversed-phase, ion-exchange, or mixed-mode retention to support a range of peptide sample-preparation strategies. The 96-well plate format can be used in centrifugation-based or automation-compatible workflows where appropriate for the selected chemistry and protocol.
Features of Thermo Scientific™ Pierce™ Peptide Clean-Up Plate, 96 well
  • High-throughput - process up to 96 peptide samples in a plate-based format
  • Flexible - choose from multiple chemistries to support plate-based peptide clean-up, desalting, concentration, and fractionation workflows
  • Efficient - help remove LC-MS-incompatible contaminants and prepare peptide samples in MS-compatible solutions for downstream analysis
  • Automation-compatible - support automated sample-preparation workflows where appropriate for the selected chemistry and protocol
Applications
  • High-throughput peptide desalting prior to liquid chromatography-mass spectrometry (LC-MS) analysis
  • Plate-based clean-up following enzymatic protein digestion workflows
  • Removal of chemistry-compatible salts, buffers, detergents, denaturants, chaotropes, lipids, and other liquid chromatography-mass spectrometry (LC-MS) interferences
  • Plate-based peptide concentration and mass spectrometry sample preparation
  • Peptide fractionation to reduce sample complexity and support deeper proteome analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 x 96-well Plate
SorbentC18
Workflow StepPeptide Clean-up
Detection MethodMass Spectrometry
FormSolid
FormatPlate
Product LinePierce
Product TypeSpin Plate
Starting MaterialPeptides, Protease-digested Protein
Unit SizeEach