What special/additional value do the KingFisher Ready MagMAX Viral/Pathogen II (MVP II) Prefilled Plates (Cat. No. A47183) provide?
The KingFisher Ready MagMAX Viral/Pathogen II (MVP II) Prefilled Plates (Cat. No. A47183) include plastics (plates, tip combs, etc.) that are otherwise purchased individually. Prefilled plate kits simplify lab inventory storage by bundling components together. They also reduce the time requirement for mid-to-high-throughput labs that otherwise have to manually pipette wash buffers and beads using standard kit reagents. Lastly, prefilled plate kits reduce hands-on interaction, thus reducing the potential for user-error and variability and the risk of re-do's and time-consuming troubleshooting.