Viral Production Medium
Viral Production Medium
Viral Production Medium
Gibco™

Viral Production Medium

Viral Production Medium is a chemically defined, serum-free, and protein-free medium for growth and transfection of Viral Production Cells 2.0.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A481790310 L
A48179011 L
A48179026 L
Catalog number A4817903
Price (USD)
2,190.65
Online exclusive
2,330.00
Save 139.35
Each
Quantity:
10 L
Price (USD)
2,190.65
Online exclusive
2,330.00
Save 139.35
Each

Gibco Viral Production Medium is a chemically defined, serum-free, and protein-free medium. It is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells).

  • For adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System
  • Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineViral Production Cells 2.0
Cell TypeHEK 293
Expression SystemMammalian
For Use With (Application)AAV Production
FormatBag
Manufacturing QualityISO 13485, MDSAP, FDA-registered
Product LineAAV-MAX
Product TypeAAV Production Medium
Quantity10 L
Shelf Life12 months from date of manufacture
Shipping ConditionRoom Temperature
ClassificationSerum-free, Animal Origin-free, Chemically-defined, Protein-free
Culture TypeSuspension Cell Culture
FormLiquid
Regulatory StatusRUO
SterilitySterile-filtered
With AdditivesGlucose, Sodium Pyruvate, Sodium Pyruvate, GlutaMAX
Without AdditivesNo GlutaMAX
pH7
Unit SizeEach
Contents & Storage
10 L bag; store at 2°C to 8°C, protected from 10 L BPC; Reagents should be stored at 2°C to 8°C, protected from light

Frequently asked questions (FAQs)

Do I need to supplement the Viral Production Medium prior to use?

Yes, prior to using the Viral Production Medium, it should be supplemented with 4 mM GlutaMAX Supplement.