TrueMark™ Comprehensive Microbiota Control
Applied Biosystems™

TrueMark™ Comprehensive Microbiota Control

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume
A503821000 μL
A5038350 μL
Catalog number A50382
Price (USD)
2,890.00
Each
Volume:
1000 μL
Price (USD)
2,890.00
Each

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research. The control can be included as a positive control for panel-specific amplification. It is available in two concentrations: 1 x 105 and 5 x 107 copies/μL. The higher concentration can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values).

NOTE: For Version 4 of TrueMark Comprehensive Microbiota Control, please contact your local representative.

The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

Note: This product will be updated on an annual basis to ensure coverage of the most popular microbial targets, and performance will be measured to ensure consistency between versions.

Version history

Version 1 of this product was first available in April 2021 and contained 471 microbial assay targets.

Version 2 became available in October 2021 and contains an additional 83 microbial assay targets.

Version 3 became avalible in March 2023 and contains an additional 139 microbial and antibiotic resistant targets.

Version 4 became available in February 2025 and contains an additional 150 microbial and antibiotic resistant targets.

Version 5 became available in September 2026 and contains an additional 86 microbial and antibiotic resistant targets.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Product LineTrueMark
Quantity1000 μL (1 x 105 copies/μL)
Shelf Life2 years
Shipping ConditionDry ice
Volume1000 μL
Concentration1 x 105 Copies/μL
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
1000 μL at a concentration of 1 x 105 copies/μL

Frequently asked questions (FAQs)

Which method/instrument was used to determine the concentration of the plasmid in the TrueMark Comprehensive Microbiota Control (Cat. No. A50382)?

The plasmid concentration was determined by means of digital PCR on the QuantStudio 3D Digital PCR System.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are the targets in the TaqMan Comprehensive Microbiota Control (Cat. No. A50382) present as single copy?

Yes, they are. However, with the exception of Xeno and RNase P which are present as multiple copies.

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.