VitroEase™ Methylamine Tungstate Negative Stain
Thermo Scientific™

VitroEase™ Methylamine Tungstate Negative Stain

Thermo Scientific VitroEase Methylamine Tungstate Negative Stain is an easy, ready-to-use negative stain for electron microscopy (EM) analysis.Features of VitroEase자세히 알아보기
Have Questions?
카탈로그 번호수량
A510365 mL
카탈로그 번호 A51036
제품 가격(KRW)
300,000
온라인 행사
Ends: 31-Dec-2026
342,000
할인액 42,000
Each
수량:
5 mL
제품 가격(KRW)
300,000
온라인 행사
Ends: 31-Dec-2026
342,000
할인액 42,000
Each
Thermo Scientific VitroEase Methylamine Tungstate Negative Stain is an easy, ready-to-use negative stain for electron microscopy (EM) analysis.

Features of VitroEase Methylamine Tungstate Negative Stain include:
• Provided as a stable, ready-to-use 2% stain solution
• Easy to use with straight-forward staining protocol
• Uniform, electron-dense stain for EM
• Dries without crystallization, ideal for EM grids

VitroEase Methylamine Tungstate Negative Stain is a ready-to-use stain with excellent spreading qualities with high density for good contrast. It wets both grid films and specimens well at neutral pH. This stain does not damage delicate structures as much as phosphotungstate (PTA) and has been found to be useful for negative staining of macromolecules, viruses, and membranes.

References:
1.Shayakhmetov, D. M.; Papayannopoulou, T.; Stamatoyannopoulos, G., and Lieber, A.: Efficient gene transfer intohuman CD34+ cells by a retargeted adenovirus vector J. Virol., 74, 2567-2583 (2000).
2. Oliver, R. M.: Negative Stain Electron Microscopy of Protein Macromolecules. Meth. Enzym., 27, 616-672 (1973).

사양
색상Colorless
등급Cryo-EM grade
물리적 형태Liquid
수량5 mL
pH6.8
Unit SizeEach