Efficient-Pro™ AGT™ Medium, 500 L - FAQs
Efficient-Pro™ AGT™ Medium, 500 L - FAQs
View additional product information for Efficient-Pro™ AGT™ Medium - FAQs (A5322301, A5322303, A5322302, A5322304)
6 product FAQs found
How should I reconstitute Efficient-Pro AGT Medium (Cat. No. A5322301, A5322302, A5322303, A5322304)?
Please follow the steps shown below to reconstitute Efficient-Pro AGT Medium (Cat. No. A5322301, A5322302, A5322303, A5322304) and to make Complete Medium:
- Measure 90% of the final volume of deionized or distilled water at room temperature (15-30 degrees C).
- Add Efficient-Pro AGT Medium at 27.3 grams/L to water.
- Mix for a minimum of 20 min.
- Use a calibrated vessel to dilute to final production volume with ambient deionized or distilled water.
- Mix for an additional 20 min.
- Measure the pH, then measure and record osmolality.
- Sterilize the product immediately by membrane filtration(positive pressure recommended).
Note: Once the product is filtered, use immediately or store at 2-8 degrees C for up to 6 months. Protect from light.
To make Complete Medium,
- Supplement Efficient-Pro AGT Medium with L‑glutamine or GlutaMAX Supplement at 2-8 mM final concentration prior to use (except for CHO-K1 GS cell lines).
Note: The recommended L‑glutamine and/or GlutaMAX Supplement concentration is 6 mM.
- Add 10 mL/L of HT Supplement for use in applications not requiring DHFR amplification.
- Glucose supplementation may be required for terminal batch cultures and should be determined empirically.
- Add Anti-Clumping Agent (1 mL/L) to the medium to reduce cell aggregation, if required.
Note: Consider reducing L‑glutamine concentration for fed-batch or perfusion protocols, or if the cell line in-use is sensitive to ammonia. Addition of a surfactant such as Pluronic™ F-68 is not required.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf-life of Efficient-Pro AGT Medium?
The shelf-life of Efficient-Pro AGT Medium is 12 months from the date of manufacture. The expiration date is provided on the product label.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
If my cell line is sensitive to ammonia, can I still use Efficient-Pro AGT Medium?
Yes, however, we recommend reducing the L-glutamine concentration if a decrease in cell viability is observed. The recommended glutamine concentration is 2-8 mM. Another way to reduced ammonia production is to use GlutaMAX Supplement instead of L-glutamine. Addition of Pluronic F-68 is not required.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does Efficient-Pro AGT Medium require supplementation for use?
When using a CHO cell line with a selection system other than the glutamine synthetase (GS) system, we recommend adding 2-8 mM L-glutamine or GlutaMAX Supplement to Efficient-Pro AGT Medium. Additionally, we recommend making sure that glucose levels do not fall below 2 g/L in culture by supplementing with glucose as needed. Also, if clumping becomes an issue, then add anti-clumping agent (ACA) to your cultures at 1% v/v.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the concentration of glucose in Efficient Pro-AGT Medium?
Efficient Pro-AGT Medium contains 4.7 g/L of glucose.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can CHO cells that are created or frozen in other production media be thawed into Efficient-Pro AGT Medium or adapted for growth and production in Efficient-Pro AGT Medium?
Cells that are grown in or adapted to, and frozen in other cell culture media readily adapt to Efficient-Pro AGT Medium. However, if your cells are experiencing a significant lag or delay in growth, please directly adapt the cells into Efficient-Pro AGT Medium and passage them for a minimum of 3 passages, until a consistent doubling time is observed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.