Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Actual product may vary
Thermo Scientific™
Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials are designed for high volume testing of residual chlorine in water samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesQuantity
AC71P2DPD Powder Refill for AQ250F2 x 250 count
AQ250FAQ250F Dispenser, 1 vial250 count
AC71P1DPD Powder Refill for AQ250F250 count
AQ250TAQ250T Dispenser, 1 vial250 count
AC72P1DPD Powder Refill for AQ250T250 count
AC72P2DPD Powder Refill for AQ250T2 x 250 count
Catalog number AC71P2
Price (USD)
119.70
Online exclusive
133.00
Save 13.30
Each
Includes:
DPD Powder Refill for AQ250F
Quantity:
2 x 250 count
Price (USD)
119.70
Online exclusive
133.00
Save 13.30
Each
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
  • Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
  • Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents 
  • Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
  • Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
  • Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
Specifications
For Use With (Equipment)AQUAfast AQ250F free chlorine DPD bulk powder dispenser
IncludesDPD Powder Refill for AQ250F
Quantity2 x 250 count
Range0.02 to 2 mg/L Cl2
Reagent FormPowder
ReagentsChlorine
Test MethodDPD
Unit SizeEach