TNF alpha Polyclonal Antibody - FAQs
TNF alpha Polyclonal Antibody - FAQs
View additional product information for TNF alpha Polyclonal Antibody - FAQs (ARC3012)
1 product FAQs found
Which antibodies directed to cytokines are suitable for use in immunohistochemistry with paraffin sections?
The following antibodies have been used successfully for IHC with paraffin sections:
Mouse (monoclonal) Anti-Human TGF-?1 (Cat. No. AHG0051), Rat (monoclonal) Anti-Human Interleukin-13 (Cat. No. AHC0132), Rabbit (polyclonal) Anti-Human IL-8 (Cat. No. AHC0881), Rabbit (polyclonal) Anti-Mouse Interferon-? (Cat. No. AMC4034), Goat (polyclonal) Anti-Mouse TNF-? (Cat. No. AMC3012), and Rabbit (polyclonal) Anti-Rat TNF-? (Cat. No. ARC3012).
You can also locate these and other antibodies with our helpful Primary Antibody Selection Guide. This can be accessed by searching "Primary Antibody Selector Tool" on our website. Once you've reached the Selection Guide, you can narrow the search by clicking on your desired links at the top of the Guide.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.