Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
B20027.06
または、製品番号B20027-06
5 g
製品番号(カタログ番号) B20027.06
または、製品番号B20027-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2905-68-2
IUPAC Namemethyl 3,4-dichlorobenzoate
Molecular FormulaC8H6Cl2O2
InChI KeyWICGATIORFSOJL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Cl)C(Cl)=C1
さらに表示
Melting Point (clear melt)44.0-53.0?C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only