Thermo Scientific Chemicals

1-Penten-3-ol, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): B20155.14
25 g, Each
Thermo Scientific Chemicals

1-Penten-3-ol, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): B20155.14
25 g, Each
数量
製品番号(カタログ番号): B20155.14
または、製品番号B20155-14

化学物質識別子

CAS
616-25-1
IUPAC Name
pent-1-en-3-ol
Molecular Formula
C5H10O
InChI Key
VHVMXWZXFBOANQ-UHFFFAOYNA-N
SMILES
CCC(O)C=C
Refractive Index
1.4220-1.4270 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

概要

1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text