Nitronium tetrafluoroborate, 96%
Nitronium tetrafluoroborate, 96%
Nitronium tetrafluoroborate, 96%
Nitronium tetrafluoroborate, 96%
Nitronium tetrafluoroborate, 96%
Thermo Scientific Chemicals

Nitronium tetrafluoroborate, 96%

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B20167.22
または、製品番号B20167-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
さらに表示
Appearance (Color)White
Assay (unspecified)>95.0% (ex Fluoride)
Formcrystals or crystalline powder
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only

General References:

  1. George A. Olah, et al. Aromatic Substitution. VIII.1 Mechanism of the Nitronium Tetrafluoroborate Nitration of Alkylbenzenes in Tetramethylene Sulfone Solution. Remarks on Certain Aspects of Electrophilic Aromatic Substitution2.J. Am. Chem. Soc.,1961,83(22), 4571-4580.
  2. George A. Olah, et al. Electrophilic reactions at single bonds. 25. Nitration of adamantane and diamantane with nitronium tetrafluoroborate.J. Am. Chem. Soc.,1993,115(6), 7246-7249.
  3. Crystalline salt of the active species in the nitration of arenes. Enables nitration to be carried out in non-aqueous, non-acidic systems (typically in Sulfolane, A13466). For mono- and dinitration of o-tolunitrile without hydrolysis, see: J. Chem. Soc., 4257 (1956); J. Am. Chem. Soc., 83, 4564 (1961).
  4. In acetonitrile solution, addition of the nitronium ion to alkenes, followed by Ritter reaction of the resulting carbonium ion with the solvent, leads to the nitroacetamide: J. Org. Chem., 36, 3641 (1971).
  5. Allylsilanes undergo desilylative nitration with allylic rearrangement; in chiral systems the reaction is enantioselective, useful in the synthesis of (E)-olefin dipeptide isosteres: J. Org. Chem., 60, 7714 (1995).
  6. Has also been used for the oxidative cleavage of O-methyl ethers of aliphatic alcohols; e.g. benzyl methyl ether is cleaved to benzaldehyde: J. Org. Chem., 42, 3097 (1977). Ethylene dithioacetals are cleaved to carbonyl compounds: Synthesis, 273 (1979).