Perfluoro-1-iodooctane, 98%
Perfluoro-1-iodooctane, 98%
Perfluoro-1-iodooctane, 98%
Perfluoro-1-iodooctane, 98%
Thermo Scientific Chemicals

Perfluoro-1-iodooctane, 98%

CAS: 507-63-1 | C8F17I | 545.965 g/mol
製品番号(カタログ番号) B20207.18
または、製品番号B20207-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS507-63-1
IUPAC Name1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
Molecular FormulaC8F17I
InChI KeyKWXGJTSJUKTDQU-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
さらに表示
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.3260-1.3300 @ 20?C
Appearance (Color)Clear colorless to pale yellow or pale purple to pale pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only