3,4-Dimethyl-2-hexanol, erythro + threo, 98%
3,4-Dimethyl-2-hexanol, erythro + threo, 98%
Thermo Scientific Chemicals

3,4-Dimethyl-2-hexanol, erythro + threo, 98%

CAS: 19550-05-1 | C8H18O | 130.231 g/mol
製品番号(カタログ番号) B20243.14
または、製品番号B20243-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19550-05-1
Refractive Index1.4320-1.4360 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5% (combination of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only