2-Ethyl-1,3-cyclopentanedione, 98%
2-Ethyl-1,3-cyclopentanedione, 98%
2-Ethyl-1,3-cyclopentanedione, 98%
Thermo Scientific Chemicals

2-Ethyl-1,3-cyclopentanedione, 98%

CAS: 823-36-9 | C7H10O2 | 126.155 g/mol
製品番号(カタログ番号) B20254.14
または、製品番号B20254-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS823-36-9
IUPAC Name2-ethylcyclopentane-1,3-dione
Molecular FormulaC7H10O2
InChI KeyYDFBIBUYOUFJMR-UHFFFAOYSA-N
SMILESCCC1C(=O)CCC1=O
さらに表示
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)168.0-178.0°C
Appearance (Color)White to cream to yellow to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only