2',4'-Dihydroxy-3'-methylacetophenone, 98%
2',4'-Dihydroxy-3'-methylacetophenone, 98%
2',4'-Dihydroxy-3'-methylacetophenone, 98%
Thermo Scientific Chemicals

2',4'-Dihydroxy-3'-methylacetophenone, 98%

CAS: 10139-84-1 | C9H10O3 | 166.176 g/mol
製品番号(カタログ番号) B20272.06
または、製品番号B20272-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10139-84-1
IUPAC Name1-(2,4-dihydroxy-3-methylphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyKMTLZBUHQPQFAV-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(O)C(C)=C1O
さらに表示
FormPowder
Appearance (Color)Dark cream to pale brown
Assay (GC)≥97.5%
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. David W. Gammon; Roger Hunter; Seanette A. Wilson. An efficient synthesis of 7-hydroxy-2,6-dimethylchromeno[3,4-d]oxazol-4-one—a protected fragment of novenamine. Tetrahedron. 2005, 61 (45), 10683-10688.
  2. J. P. Brown; D. H. Johnson; Alexander Robertson and W. B. Whalley. 448. Some alkylresorcinols. J. Chem. Soc. 1951, 2019-2020.