3-Amidinopyridine hydrochloride, 97%
3-Amidinopyridine hydrochloride, 97%
3-Amidinopyridine hydrochloride, 97%
Thermo Scientific Chemicals

3-Amidinopyridine hydrochloride, 97%

CAS: 7356-60-7 | C6H8ClN3 | 157.601 g/mol
製品番号(カタログ番号) B20406.03
または、製品番号B20406-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7356-60-7
IUPAC Namepyridine-3-carboximidamide hydrochloridyl
Molecular FormulaC6H7ClN3
InChI KeySCYPVJVMZZPCFF-UHFFFAOYSA-N
SMILES[Cl].NC(=N)C1=CC=CN=C1
さらに表示
Appearance (Color)White
Melting Point (clear melt)186.0-196.0?C
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only