2-Methoxyphenoxyacetic acid, 98+%
2-Methoxyphenoxyacetic acid, 98+%
2-Methoxyphenoxyacetic acid, 98+%
Thermo Scientific Chemicals

2-Methoxyphenoxyacetic acid, 98+%

CAS: 1878-85-9 | C9H10O4 | 182.175 g/mol
製品番号(カタログ番号) B20672.14
または、製品番号B20672-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1878-85-9
IUPAC Name2-(2-methoxyphenoxy)acetic acid
Molecular FormulaC9H10O4
InChI KeyIHONYPFTXGQWAX-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1OCC(O)=O
さらに表示
FormPowder or needles
Appearance (Color)White to pale brown or pale grey
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)118.0-126.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only