Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals
Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals
Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals
Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals

数量:
25 g
100 g
500 g
製品番号(カタログ番号) B20759.14
または、製品番号B20759-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS145224-92-6
IUPAC Namesodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate hydrate
Molecular FormulaC24H41NaO5
InChI KeyNDVHNZISGVSFMP-WTCAICSISA-M
SMILESO.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O
さらに表示
Appearance (Color)White
Optical Rotation44.0 ± 2.0° (C=2 in water, UK-sourced material)
FormPowder
Assay from Supplier's CofA≥97.5% (HPLC, US-sourced material)
Identification (FTIR)Conforms (UK-sourced material)
さらに表示
Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Christie, M.; Torres, R. M.; Kedl, R. M.; Randolph, T. W.; Carpenter, J. F. Recombinant murine growth hormone particles are more immunogenic with intravenous than subcutaneous administration. J. Pharm. Sci. 2014, 103 (1), 128-139.
  2. Kuhre, R. E.; Albrechtsen, N. W.; Windeløv, J. A.; Svendsen, B.; Hartmann, B.; Holst, J. J. GLP-1 amidation efficiency along the length of the intestine in mice, rats and pigs and in GLP-1 secreting cell lines. Peptides 2014, 55, 52-57.