2,3-Dichlorophenyl isothiocyanate, 97%
2,3-Dichlorophenyl isothiocyanate, 97%
2,3-Dichlorophenyl isothiocyanate, 97%
Thermo Scientific Chemicals

2,3-Dichlorophenyl isothiocyanate, 97%

CAS: 6590-97-2 | C7H3Cl2NS | 204.07 g/mol
製品番号(カタログ番号) B20930.06
または、製品番号B20930-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6590-97-2
IUPAC Name1,2-dichloro-3-isothiocyanatobenzene
Molecular FormulaC7H3Cl2NS
InChI KeyRQZIODPVCCTBAQ-UHFFFAOYSA-N
SMILESClC1=CC=CC(N=C=S)=C1Cl
さらに表示
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.6725-1.6775 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only