Thermo Scientific Chemicals

4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B20989.06
5 g, Each
Thermo Scientific Chemicals

4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B20989.06
5 g, Each
Quantity
Catalog number: B20989.06
also known as B20989-06
Price (USD)
Price: 69.10
Online price: 58.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15687-27-1
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
75-79?C
Appearance (Color)
White
Assay (HPLC)
≥98.5%
Form
Crystals or powder or crystalline powder

Description

Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text