Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%

CAS: 50551-58-1 | C12H12O4 | 220.22 g/mol
製品番号(カタログ番号) B21066.14
または、製品番号B21066-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS50551-58-1
IUPAC Nameethyl 7-methoxy-1-benzofuran-2-carboxylate
Molecular FormulaC12H12O4
InChI KeySXCMZYVXJUJIDJ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC=CC(OC)=C2O1
さらに表示
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)83.0-93.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only