Thermo Scientific Chemicals

D-Aspartic acid, 99%

Catalog number: B21184.22
100 g, Each
Thermo Scientific Chemicals

D-Aspartic acid, 99%

Catalog number: B21184.22
100 g, Each
Quantity
Catalog number: B21184.22
also known as B21184-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1783-96-6
IUPAC Name
2-aminobutanedioic acid
Molecular Formula
C4H7NO4
InChI Key
CKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILES
NC(CC(O)=O)C(O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Optical Rotation
-25.0° ± 0.5° (c=1 in 1N HCl)
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

D-Aspartic acid modulates melatonin synthesis in the pineal gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid modulates melatonin synthesis in the pineal gland.

Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text