Pseudothiohydantoin, 97%
Pseudothiohydantoin, 97%
Pseudothiohydantoin, 97%
Thermo Scientific Chemicals

Pseudothiohydantoin, 97%

CAS: 556-90-1 | C3H4N2OS | 116.138 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B21461.06
または、製品番号B21461-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS556-90-1
IUPAC Name2-amino-4,5-dihydro-1,3-thiazol-4-one
Molecular FormulaC3H4N2OS
InChI KeyHYMJHROUVPWYNQ-UHFFFAOYSA-N
SMILESNC1=NC(=O)CS1
さらに表示
Appearance (Color)White to cream
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only