Thermo Scientific Chemicals

Pseudothiohydantoin, 97%, Thermo Scientific Chemicals

Catalog number: B21461.14
25 g, Each
Thermo Scientific Chemicals

Pseudothiohydantoin, 97%, Thermo Scientific Chemicals

Catalog number: B21461.14
25 g, Each
Quantity
Catalog number: B21461.14
Price (USD)
Price: 125.00
Online price: 108.65
Your price:
Quantity
-

Chemical Identifiers

CAS
556-90-1
IUPAC Name
2-amino-4,5-dihydro-1,3-thiazol-4-one
Molecular Formula
C3H4N2OS
InChI Key
HYMJHROUVPWYNQ-UHFFFAOYSA-N
SMILES
NC1=NC(=O)CS1
Appearance (Color)
White to cream
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text