Thermo Scientific Chemicals

Tolbutamide, 98%, Thermo Scientific Chemicals

Catalog number: B21698.30
250 g, Each
Thermo Scientific Chemicals

Tolbutamide, 98%, Thermo Scientific Chemicals

Catalog number: B21698.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21698.30
also known as B21698-30
Price (USD)
Price: 632.00
Online price: 546.65
Your price:
Quantity
-

Chemical Identifiers

CAS
64-77-7
IUPAC Name
3-butyl-1-(4-methylbenzenesulfonyl)urea
Molecular Formula
C12H18N2O3S
InChI Key
JLRGJRBPOGGCBT-UHFFFAOYSA-N
SMILES
CCCCNC(=O)NS(=O)(=O)C1=CC=C(C)C=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (UK-sourced material)
Assay (HPLC)
≥97.5%
Comment
Material sourced in UK and US
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Melting Point (clear melt)
126-130°C

Description

Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text