N(epsilon)-Boc-L-lysine, 97%
N(epsilon)-Boc-L-lysine, 97%
N(epsilon)-Boc-L-lysine, 97%
Thermo Scientific Chemicals

N(epsilon)-Boc-L-lysine, 97%

CAS: 2418-95-3 | C11H22N2O4 | 246.31 g/mol
製品番号(カタログ番号) B21738.03
または、製品番号B21738-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2418-95-3
IUPAC Name(2S)-2-azaniumyl-6-{[(tert-butoxy)carbonyl]amino}hexanoate
Molecular FormulaC11H22N2O4
InChI KeyVVQIIIAZJXTLRE-QMMMGPOBSA-N
SMILESCC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O
さらに表示
FormPowder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Optical Rotation17° ± 2° (c=1 in acetic acid)
It is used as therapy to treat recurrent herpes simplex infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as therapy to treat recurrent herpes simplex infections.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Takahito Mukai et. al. Addingl-lysine derivatives to the genetic code of mammalian cells with engineered pyrrolysyl-tRNA synthetases. Biochemical and Biophysical Research Communications, 2008, 371 (4), 818-822.
  2. K Itakura; and K Uchida.Evidence that malondialdehyde-derived aminoenimine is not a fluorescent age pigment. Chemical Research in Toxicology, 2001, 14 (5), 473-475.