Thermo Scientific Chemicals

5-Fluorosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21751.06
5 g, Each
Thermo Scientific Chemicals

5-Fluorosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21751.06
5 g, Each
Quantity
Catalog number: B21751.06
also known as B21751-06
Price (USD)
Price: 222.00
Online price: 191.65
Your price:
Quantity
-

Chemical Identifiers

CAS
345-16-4
IUPAC Name
5-fluoro-2-hydroxybenzoic acid
Molecular Formula
C7H5FO3
InChI Key
JWPRICQKUNODPZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(F)=CC=C1O
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥96.0%
Appearance (Color)
White
Melting Point (clear melt)
175.0-185.0?C
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text