Thermo Scientific Chemicals

2-Bromobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: B21840.03
1 g, Each
Thermo Scientific Chemicals

2-Bromobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: B21840.03
1 g, Each
Quantity
Catalog number: B21840.03
also known as B21840-03
Price (USD)

Chemical Identifiers

CAS
2052-07-5
IUPAC Name
2-bromo-1,1'-biphenyl
Molecular Formula
C12H9Br
InChI Key
KTADSLDAUJLZGL-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=C1C1=CC=CC=C1
Appearance (Color)
Clear, colorless to yellow
Form
Liquid
Assay (GC)
>97.5%

Description

It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text