1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
Thermo Scientific Chemicals

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%

CAS: 52431-78-4 | C7H6N4OS | 194.212 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B22003.14
または、製品番号B22003-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS52431-78-4
IUPAC Name1-(4-hydroxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Molecular FormulaC7H6N4OS
InChI KeyMOXZSKYLLSPATM-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)N1NN=NC1=S
さらに表示
FormCrystals, crystalline powder or pieces
Assay (HPLC)≥95.0%
Loss on Drying≤5.0% (105?C/constant weight)
Solution Test25 mg/ml in Methanol: clear colorless to pale yellow or pink to purple solution
Appearance (Color)White to cream to purple
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only