2,4-Dinitrophenylacetic acid, 98%
2,4-Dinitrophenylacetic acid, 98%
2,4-Dinitrophenylacetic acid, 98%
2,4-Dinitrophenylacetic acid, 98%
Thermo Scientific Chemicals

2,4-Dinitrophenylacetic acid, 98%

CAS: 643-43-6 | C8H6N2O6 | 226.144 g/mol
製品番号(カタログ番号) B22102.14
または、製品番号B22102-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS643-43-6
IUPAC Name2-(2,4-dinitrophenyl)acetate
Molecular FormulaC8H5N2O6
InChI KeyKCNISYPADDTFDO-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
さらに表示
Appearance (Color)Pale cream to pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only