4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
4-(Hydroxymethyl)benzoic acid, 98+%
Thermo Scientific Chemicals

4-(Hydroxymethyl)benzoic acid, 98+%

CAS: 3006-96-0 | C8H8O3 | 152.15 g/mol
製品番号(カタログ番号) B22183.06
または、製品番号B22183-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3006-96-0
IUPAC Name4-(hydroxymethyl)benzoic acid
Molecular FormulaC8H8O3
InChI KeyWWYFPDXEIFBNKE-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C(O)=O
さらに表示
Appearance (Color)White to pale cream to gray
Assay (HPLC)≥98.0%
FormPowder or flakes
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)178.0-184.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only