N-Phenylglycine ethyl ester, 99%
N-Phenylglycine ethyl ester, 99%
N-Phenylglycine ethyl ester, 99%
Thermo Scientific Chemicals

N-Phenylglycine ethyl ester, 99%

CAS: 2216-92-4 | C10H13NO2 | 179.219 g/mol
製品番号(カタログ番号) B22282.09
または、製品番号B22282-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2216-92-4
IUPAC Nameethyl 2-(phenylamino)acetate
Molecular FormulaC10H13NO2
InChI KeyMLSGRWDEDYJNER-UHFFFAOYSA-N
SMILESCCOC(=O)CNC1=CC=CC=C1
さらに表示
Assay (GC)≥98.5%
Appearance (Color)Cream to pale brown
Melting Point (clear melt)54.5-58.5?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only