trans-2-Aminocyclohexanol hydrochloride, 97+%
trans-2-Aminocyclohexanol hydrochloride, 97+%
trans-2-Aminocyclohexanol hydrochloride, 97+%
Thermo Scientific Chemicals

trans-2-Aminocyclohexanol hydrochloride, 97+%

CAS: 5456-63-3 | C6H13ClNO | 150.63 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B22324.06
または、製品番号B22324-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5456-63-3
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or granules
Assay (Titration ex Chloride)≥97.0 to ≤103.0% (Dry wt. basis)
Water Content (Karl Fischer Titration)≤2.0%
Melting Point (clear melt)168.0-179.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only