1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents, acids.
RUO – Research Use Only
General References:
- Farzana Aslam; David J. Binks; Mark D. Rahn; David P. West; Paul O'Brien & Nigel Pickett. Photorefractive performance of polymer composite sensitized by CdSe nanoparticles passivated by 1-hexadecylamine. Journal of Modern Optics. 2003, 52 (7), 945-953.
- Xiaomiao Hou; Xiaoling Zhang; Yan Fang; Shutang Chen; Na Li; Qi Zhou. 1-Hexadecylamine as both reducing agent and stabilizer to synthesize Au and Ag nanoparticles and their SERS application. Journal of Nanoparticle Research. 2011, 13 (5), 1929-1936.