Thermo Scientific Chemicals

o-Thiocresol, 98%, Thermo Scientific Chemicals

Catalog number: B22460.14
25 g, Each
Thermo Scientific Chemicals

o-Thiocresol, 98%, Thermo Scientific Chemicals

Catalog number: B22460.14
25 g, Each
Quantity
Catalog number: B22460.14
also known as B22460-14
Price (USD)
Price: 127.00
Online price: 109.65
Your price:
Quantity
-

Chemical Identifiers

CAS
137-06-4
IUPAC Name
2-methylbenzene-1-thiol
Molecular Formula
C7H8S
InChI Key
LXUNZSDDXMPKLP-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1S
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5790-1.5830 @ 20?C

Description

o-Thiocresol is used as a thiol for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Thiocresol is used as a thiol for proteomics research.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text