Thermo Scientific Chemicals

5-Bromo-2-furaldehyde, 98%, stab. with 2% ethanol, Thermo Scientific Chemicals

Catalog number: B22536.18
50 g, Each
Thermo Scientific Chemicals

5-Bromo-2-furaldehyde, 98%, stab. with 2% ethanol, Thermo Scientific Chemicals

Catalog number: B22536.18
50 g, Each
Quantity
Catalog number: B22536.18
also known as B22536-18
Price (USD)
Price: 345.00
Online price: 295.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1899-24-7
IUPAC Name
5-bromofuran-2-carbaldehyde
Molecular Formula
C5H3BrO2
InChI Key
WJTFHWXMITZNHS-UHFFFAOYSA-N
SMILES
BrC1=CC=C(O1)C=O
Form
Crystalline powder
Assay (GC)
>97.5%
Free acid (titration)
<1.0%
Appearance (Color)
White to pale cream/yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text