Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals
Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals
Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals
Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号) B22559.06
または、製品番号B22559-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6779-09-5
IUPAC Nameethylphosphonate
Molecular FormulaC2H5O3P
InChI KeyGATNOFPXSDHULC-UHFFFAOYSA-L
SMILESCCP([O-])([O-])=O
さらに表示
Appearance (Color)White to off-white
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)57.0-66.0°C
Assay from Suppliers CofA≥98.0%
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Parker, S. L.; Roberts, J. A.; Lipman, J.; Wallis, S. C. Quantitative bioanalytical validation of fosfomycin in human whole blood with volumetric absorptive microsampling. Bioanalysis 2015, 7 (19), 2585-2595.
  2. Pauly, C. S.; Genix, A. C.; Alauzun, J. G.; Sztucki, M.; Oberdisse, J.; Mutin, P. H. Surface modification of alumina-coated silica nanoparticles in aqueous sols with phosphonic acids and impact on nanoparticle interactions. Phys. Chem. Chem. Phys. 2015, 17 (29), 19173-19182.