2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%
2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%
2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%
Thermo Scientific Chemicals

2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%

CAS: 6813-38-3 | C12H6N2O4 | 242.19 g/mol
製品番号(カタログ番号) B22594.03
または、製品番号B22594-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6813-38-3
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or gray or pale pink
Sulfated Ash≤3.0%
Assay (Silylated GC)≥97.5%
2,2'-Bipyridine-4,4'-dicarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Md. K.Nazeeruddin; S.M.Zakeeruddin; R.Humphry-Baker; M.Jirousek; P.Liska; N.Vlachopoulos; V.Shklover; Christian-H. Fischer; M.Gratzel. Acid-base equilibria of (2,2‘-bipyridyl-4,4‘-dicarboxylic acid)ruthenium(II) complexes and the effect of protonation on charge-transfer sensitization of nanocrystalline titania. Inorg. Chem. 1999, 38,(26), 6298-6305. 
  2. Susan Anderson; Edwin C.Constable; Kenneth R.Seddon; Janet E.Turp; James E.Baggott; Michael J.Pilling. Preparation and characterisation of 2,2'-bipyridine-4,4'-disulphonic and -5-sulphonic acids and their ruthenium(II) complexes. Excited-state properties and excited-state electron-transfer reactions of ruthenium(II) complexes containing 2,2'-bipyridine-4,4'-disulphonic acid or 2,2'-bipyridine-4,4' dicarboxylic acid. J. Chem. Soc., Dalton Trans. 1985, (11), 2247-2261.