2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号) B22662.22
または、製品番号B22662-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4261-68-1
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1%
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)130.0-136.0?C
Appearance (Color)Pale cream to cream
さらに表示
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Forsyth, S. A.; Fröhlich, U.; Goodrich, P.; Gunaratne, H. N.; Hardacre, C.; McKeown, A.; Seddon, K. R. Functionalised ionic liquids: synthesis of ionic liquids with tethered basic groups and their use in Heck and Knoevenagel reactions. New J. Chem. 2010, 34 (4), 723-731.
  2. Richards, J. M.; McClennen, W. H.; Meuzelaar, H. L. Determination of additives in biomer and lycra spandex by pyrolysis tandem mass spectrometry and time temperature resolved pyrolysis mass spectrometry. J. Appl. Polym. Sci. 1990, 40 (1-2), 1-12.