Thermo Scientific Chemicals

3-Chlorothiophene, 98%, may contain up to 2% DMF, Thermo Scientific Chemicals

Catalog number: B22669.14
25 g, Each
Thermo Scientific Chemicals

3-Chlorothiophene, 98%, may contain up to 2% DMF, Thermo Scientific Chemicals

Catalog number: B22669.14
25 g, Each
Quantity
Catalog number: B22669.14
Price (USD)
Price: 150.00
Online price: 129.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17249-80-8
IUPAC Name
3-chlorothiophene
Molecular Formula
C4H3ClS
InChI Key
QUBJDMPBDURTJT-UHFFFAOYSA-N
SMILES
ClC1=CSC=C1
Comment
DMF remains from the manufacturing process
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5% (excluding DMF)
Refractive Index
1.5480-1.5540 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text