4-Phenyl-2-butanone, 98%
4-Phenyl-2-butanone, 98%
4-Phenyl-2-butanone, 98%
Thermo Scientific Chemicals

4-Phenyl-2-butanone, 98%

CAS: 2550-26-7 | C10H12O | 148.205 g/mol
製品番号(カタログ番号) B22777.0B
または、製品番号B22777-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2550-26-7
IUPAC Name4-phenylbutan-2-one
Molecular FormulaC10H12O
InChI KeyAKGGYBADQZYZPD-UHFFFAOYSA-N
SMILESCC(=O)CCC1=CC=CC=C1
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5105-1.5145 @ 20?C
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Melvin S.Newman; Gideon Fraenkel; Walter N.Kirn. The reactions of 4-phenyl-2-butanone and 5-phenyl-2-pentanone with phosphorus pentachloride1. J. Org. Chem. 1963, 28,(7), 1851-1853.
  2. John B.Melpolder; Richard F.Heck. Palladium-catalyzed arylation of allylic alcohols with aryl halides. J. Org. Chem. 1976, 41,(2), 265-272.