Phenyltrimethylammonium chloride, 98+%
Phenyltrimethylammonium chloride, 98+%
Phenyltrimethylammonium chloride, 98+%
Thermo Scientific Chemicals

Phenyltrimethylammonium chloride, 98+%

CAS: 138-24-9 | C9H14ClN | 171.67 g/mol
製品番号(カタログ番号) B22851.22
または、製品番号B22851-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS138-24-9
IUPAC NameN,N,N-trimethylanilinium chloride
Molecular FormulaC9H14ClN
InChI KeyMQAYPFVXSPHGJM-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)C1=CC=CC=C1
さらに表示
Appearance (Color)White
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.0%
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

Solubility
Soluble in water 333 g/L.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
RUO – Research Use Only

General References:

  1. Paul G. Sears; Eugene D. Wilhoit and Lyle R. Dawson. Conductances of Trimethylphenylammonium Chloride and Iodide in Water and in Dimethylformamide at 25°C. J. Chem. Phys. 1955, 23 (7), 1274.
  2. T. Jiang; M.J. Chollier Brym; G. Dubéb, A. Lasia; G.M. Brisard. Electrodeposition of Aluminum from ionic liquids: Part II - studies on the electrodeposition of aluminum from aluminum chloride (AICl3) - trimethylphenylammonium chloride (TMPAC) ionic liquids. Surface and Coatings Technology. 2006, 201, (1-2), 10-18.