2-Chloro-4-fluorophenylacetic acid, 97%
2-Chloro-4-fluorophenylacetic acid, 97%
2-Chloro-4-fluorophenylacetic acid, 97%
2-Chloro-4-fluorophenylacetic acid, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluorophenylacetic acid, 97%

CAS: 177985-32-9 | C8H6ClFO2 | 188.582 g/mol
製品番号(カタログ番号) B22872.06
または、製品番号B22872-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS177985-32-9
IUPAC Name2-(2-chloro-4-fluorophenyl)acetate
Molecular FormulaC8H5ClFO2
InChI KeyCDUSPKFHAVQVRQ-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=C(F)C=C1Cl
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)98.5-107.5°C
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Aydın Alemdar; Ali Rıza Özkaya; Mustafa Bulut. Preparation, characterization, electrochemistry and in situ spectroelectrochemistry of novel α-tetra[7-oxo-3-(2-chloro-4-fluorophenyl)coumarin]-substituted metal-free, cobalt and zinc phthalocyanines. Synthetic Metals. 2010, 160 (13-14), 1556-1565.
  2. Aydın Alemdar; Ali Rıza Özkaya; Mustafa Bulut. Synthesis, spectroscopy, electrochemistry and in situ spectroelectrochemistry of partly halogenated coumarin phthalonitrile and corresponding metal-free, cobalt and zinc phthalocyanines. Polyhedron. 2009, 28 (17), 3788-3796.