Thermo Scientific Chemicals

Methoxyacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22917.18
50 g, Each
Thermo Scientific Chemicals

Methoxyacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22917.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B22917.18
also known as B22917-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
625-45-6
IUPAC Name
2-methoxyacetic acid
Molecular Formula
C3H6O3
InChI Key
RMIODHQZRUFFFF-UHFFFAOYSA-N
SMILES
COCC(O)=O
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4140-1.4190 @ 20?C
Appearance (Color)
Clear colorless

Description

Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text