1,4-Cyclohexanedicarboxylic acid, cis + trans, 98%
1,4-Cyclohexanedicarboxylic acid, cis + trans, 98%
1,4-Cyclohexanedicarboxylic acid, cis + trans, 98%
Thermo Scientific Chemicals

1,4-Cyclohexanedicarboxylic acid, cis + trans, 98%

CAS: 1076-97-7 | C8H12O4 | 172.18 g/mol
製品番号(カタログ番号) B23125.36
または、製品番号B23125-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1076-97-7
IUPAC Namecyclohexane-1,4-dicarboxylic acid
Molecular FormulaC8H12O4
InChI KeyPXGZQGDTEZPERC-UHFFFAOYSA-N
SMILESOC(=O)C1CCC(CC1)C(O)=O
さらに表示
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only