3-Bromopropylamine hydrobromide, 98%
3-Bromopropylamine hydrobromide, 98%
3-Bromopropylamine hydrobromide, 98%
Thermo Scientific Chemicals

3-Bromopropylamine hydrobromide, 98%

CAS: 5003-71-4 | C3H9Br2N | 218.92 g/mol
製品番号(カタログ番号) B23254.22
または、製品番号B23254-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5003-71-4
IUPAC Namehydrogen 3-bromopropan-1-amine bromide
Molecular FormulaC3H9Br2N
InChI KeyPQIYSSSTRHVOBW-UHFFFAOYSA-N
SMILES[H+].[Br-].NCCCBr
さらに表示
Appearance (Color)White to pale cream to pale pink
FormPowder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.

Solubility
Soluble in water (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only

General References:

  1. Eleanor D. Bates; Rebecca D. Mayton; Ioanna Ntai and James H. Davis , Jr. CO2 Capture by a Task-Specific Ionic Liquid. J. Am. Chem. Soc. 2002, 124 (6), 926-927.
  2. R. F. Parcell; F. P. HauckJr. The Preparation of Tetrahydropyridines from Enamines and Imines. J. Org. Chem. 1963, 28 (12), 3468-3473.