2-Ethylhexyl acetate, 99%
2-Ethylhexyl acetate, 99%
2-Ethylhexyl acetate, 99%
Thermo Scientific Chemicals

2-Ethylhexyl acetate, 99%

CAS: 103-09-3 | C10H20O2 | 172.27 g/mol
数量:
250 mL
1000 mL
製品番号(カタログ番号) B23257.AK
または、製品番号B23257-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS103-09-3
IUPAC Name2-ethylhexyl acetate
Molecular FormulaC10H20O2
InChI KeyWOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(C)=O
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only